missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-hydroxy-5-nitrophenol)disulfide Disodium Salt, TRC

Product Code. 30336403
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336403 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30336403 Supplier Toronto Research Chemicals Supplier No. TRCB44675010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis(2-hydroxy-5-nitrophenol)disulfide Disodium Salt
Molecular Formula C12 H6 N2 Na2 O6 S2
Quantity 10 g
Synonym 2,2'-Dithiobis[4-nitro-phenol Disodium Salt
InChI Formula InChI=1S/C12H8N2O6S2.2Na/c15-9-3-1-7(13(17)18)5-11(9)21-22-12-6-8(14(19)20)2-4-10(12)16;;/h1-6,15-16H;;/q;2*+1/p-2
SMILES [O-][N+](=O)c1ccc(O[Na])c(SSc2cc(ccc2O[Na])[N+](=O)[O-])c1
IUPAC Name [4-nitro-2-[(5-nitro-2-sodiooxyphenyl)disulfanyl]phenoxy]sodium
Molecular Weight (g/mol) 384.295
Formula Weight 383.946
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.