Learn More
Bis(2-chloroethyl)-1,1,2,2-d4-amine Hydrochloride, TRC
Product Code. 30347977
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30347977 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30347977 Supplier Toronto Research Chemicals Supplier No. TRCB418803100MG
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket
Add to basket
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Bis(2-chloroethyl)-1,1,2,2-d4-amine hydrochloride
CAS 352431-06-2
Molecular Formula C4 2H4 H5 Cl2 N . Cl H
Quantity 100 mg
Synonym Ethan-1,1,2,2-d4-amine, 2-chloro-N-(2-chloroethyl)-, hydrochloride (9CI), Bis(2-chloroethyl)-1,1,2,2-tetradeuterioamine hydrochloride
InChI Formula InChI=1S/C4H9Cl2N.ClH/c5-1-3-7-4-2-6;/h7H,1-4H2;1H/i1D2,3D2;
SMILES Cl.[2H]C([2H])(Cl)C([2H])([2H])NCCCl
IUPAC Name 2-chloro-N-(2-chloroethyl)-1,1,2,2-tetradeuterioethanamine;hydrochloride
Molecular Weight (g/mol) 182.51
Formula Weight 181.013