Learn More
Bis(2-butoxyethyl) 2-Hydroxyethyl-d4 Phosphate Triester, TRC
Product Code. 30336436
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
Unit Size:
1mg
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336436 1 mg 1mg
30330012 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30336436 Supplier Toronto Research Chemicals Supplier No. TRCB4151921MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Bis(2-butoxyethyl) 2-Hydroxyethyl-d4 Phosphate Triester
Molecular Formula C14 D4 H27 O7 P
Quantity 1 mg
Synonym Bis(2-butoxyethyl) 2-Hydroxyethyl Ester Phosphoric Acid-d4
InChI Formula InChI=1S/C14H31O7P/c1-3-5-8-17-11-13-20-22(16,19-10-7-15)21-14-12-18-9-6-4-2/h15H,3-14H2,1-2H3/i7D2,10D2
SMILES [2H]C([2H])(O)C([2H])([2H])OP(=O)(OCCOCCCC)OCCOCCCC
IUPAC Name bis(2-butoxyethyl) (1,1,2,2-tetradeuterio-2-hydroxy-ethyl) phosphate
Molecular Weight (g/mol) 346.39
Formula Weight 346.2058