Learn More
Bis(2-butoxyethyl) 2-Hydroxyethyl-d4 Phosphate Triester, TRC
Specifications
Specifications
| Chemical Name or Material | Bis(2-butoxyethyl) 2-Hydroxyethyl-d4 Phosphate Triester |
| Molecular Formula | C14 D4 H27 O7 P |
| Quantity | 2.5 mg |
| Synonym | Bis(2-butoxyethyl) 2-Hydroxyethyl Ester Phosphoric Acid-d4 |
| InChI Formula | InChI=1S/C14H31O7P/c1-3-5-8-17-11-13-20-22(16,19-10-7-15)21-14-12-18-9-6-4-2/h15H,3-14H2,1-2H3/i7D2,10D2 |
| SMILES | [2H]C([2H])(O)C([2H])([2H])OP(=O)(OCCOCCCC)OCCOCCCC |
| IUPAC Name | bis(2-butoxyethyl) (1,1,2,2-tetradeuterio-2-hydroxy-ethyl) phosphate |
| Molecular Weight (g/mol) | 346.39 |
| Formula Weight | 346.2058 |