missing translation for 'onlineSavingsMsg'
Learn More

Bis(2-benzyloxy-3-nitrophenyl)disulfide, TRC

Product Code. 30348134
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30348134 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348134 Supplier Toronto Research Chemicals Supplier No. TRCB410950500MG
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bis(2-benzyloxy-3-nitrophenyl)disulfide
CAS 37398-25-7
Molecular Formula C26 H20 N2 O6 S2
Quantity 500 mg
InChI Formula InChI=1S/C26H20N2O6S2/c29-27(30)21-13-7-15-23(25(21)33-17-19-9-3-1-4-10-19)35-36-24-16-8-14-22(28(31)32)26(24)34-18-20-11-5-2-6-12-20/h1-16H,17-18H2
SMILES [O-][N+](=O)c1cccc(SSc2cccc(c2OCc3ccccc3)[N+](=O)[O-])c1OCc4ccccc4
IUPAC Name 1-nitro-3-[(3-nitro-2-phenylmethoxyphenyl)disulfanyl]-2-phenylmethoxybenzene
Molecular Weight (g/mol) 520.58
Formula Weight 520.0763
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.