Learn More
Bis(2,5-dioxopyrrolidin-1-yl) succinate, TRC
Specifications
Specifications
| Chemical Name or Material | Bis(2,5-dioxopyrrolidin-1-yl) succinate |
| CAS | 30364-60-4 |
| Molecular Formula | C12H12N2O8 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C12H12N2O8/c15-7-1-2-8(16)13(7)21-11(19)5-6-12(20)22-14-9(17)3-4-10(14)18/h1-6H2 |
| SMILES | O=C(CCC(=O)ON1C(=O)CCC1=O)ON2C(=O)CCC2=O |
| IUPAC Name | bis(2,5-dioxopyrrolidin-1-yl) butanedioate |
| Molecular Weight (g/mol) | 312.23 |
| Formula Weight | 312.0594 |