missing translation for 'onlineSavingsMsg'
Learn More

Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II), 99.9% (metals basis)

Product Code. 11319667
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11319667

Brand: Thermo Scientific Alfa Aesar 041279.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

CAS: 14040-05-2 | C22H38CuO4 | 430.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14040-05-2
Molecular Formula C22H38CuO4
Molecular Weight (g/mol) 430.09
MDL Number MFCD00058920
InChI Key JGMGCTZGOAONPQ-UHFFFAOYSA-N
Synonym copper bis 2,2,6,6-tetramethyl-3,5-heptanedionate, 2,2,6,6-tetramethyl-5-2,2,6,6-tetramethyl-5-oxohept-3-en-3-yl oxy cuprio oxy hept-4-en-3-one
PubChem CID 91663146
SMILES [Cu++].CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C.CC(C)(C)C(=O)[CH-]C(=O)C(C)(C)C

Specifications

Melting Point 198°C
Boiling Point 100°C (0.1 mmHg)(sublimation)
Quantity 25 g
IUPAC Name copper;5-hydroxy-2,2,6,6-tetramethylhept-4-en-3-one
Formula Weight 430.05
Percent Purity 99.9%
Physical Form Powder
Assay (metals basis)
Chemical Name or Material Bis(2,2,6,6-tetramethyl-3,5-heptanedionato)copper(II)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.