Learn More
Bis(1,5-cyclooctadiene)iridium (I) tetrafluoroborate, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 35138-23-9 |
| Molecular Formula | C16H24BF4Ir |
| MDL Number | MFCD00079664 |
| Quantity | 250 mg |
| SMILES | F[B-](F)(F)F.[Ir+].C1=C\CC/C=C\CC/1.C2=C\CC/C=C\CC/2 |
| Molecular Weight (g/mol) | 495.38 |
| Percent Purity | 97% |
| Chemical Name or Material | Bis(1,5-cyclooctadiene)iridium (I) tetrafluoroborate |