missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Application Notes:
- Biotin-X-Val-Ala-Asp(OMe)-FMK is biotinylated derivative of Z-VAD(OMe)-FMK that may be used to identify active caspases in cell lysates.
Usage Statement:
- Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Key Applications:
- Caspase inhibitor/Probe
Specifications
Specifications
| Product Type | Biotin Tag Caspase Inhibitor |
| Form | Solid |
| For Use With (Application) | Caspase Inhibitor |
| Sufficient For | Identifying Active Caspases in Cell Lysates |
| Quantity | 1 mg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?