Learn More
Thermo Scientific Chemicals Biotin-Substance P
Product Code. 15448099
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 15448099
Brand: Thermo Scientific Chemicals J66653.MA
Request Bulk or Custom Format
For cellular and molecular biology applications
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Purification Method HPLC
Content And Storage -30°C to -10°C
Format Solid
Molecular Weight (g/mol) 1573.94
Product Type Biotin-Substance P
Quantity 10 mg
Regulatory Status RUO - research use only
Sequence Biotin-Arg-Pro-Lys-Pro-Gln-Gln-Phe-Phe-Gly-Leu-Met-NH2