missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Biosate™ Peptone

Product Code. 16249761
Change view
Click to view available options
Quantity:
454 g
Unit Size:
454g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16249761 454 g 454g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16249761 Supplier Gibco™ Supplier No. 211862
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy

An animal origin (AO) mixed hydrolysate comprised of 65% pancreatic digest of casein and 35% yeast extract.

The combination of pancreatic digest of casein and yeast extract provides synergistic nutritional benefits that are not provided by the components alone, resulting in a unique nutritional profile. Customers have shown the combined use of these two peptones, when supplied as Biosate Peptone, improves toxin production with Clostridial species for vaccines, such as Clostridium botulinum and Clostridium perfringens.
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance LUO, MUO
Content And Storage Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Form Powder
Product Type Dehydrated Culture Media
Format Bottle
For Use With (Application) Nutritional supplement in vitamin assays, susceptibility testing, and other laboratory media, and microbial fermentation in which the high salt content will not interfere
Product Line Biosate
Quantity 454 g
Source Animal origin, casein and yeast
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.