Learn More
Sartorius™ Biohit™ Proline Single-Channel Mechanical Pipettes
Shop All Sartorius Products
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
0.2 to 2.5 μL
0.5 to 10 μL
2 to 20 μL
5 to 50 μL
missing translation for 'unitSize'
Each
Beschreibung
- Light weight with high accuracy and precision for use by professionals
- Wide range of pipettes with adjustable or fixed volume ranges
- Easy volume setting with click stop mechanism
- Relaxed grip with ergonomic finger support
- Reduced risk of contamination with Safe-Cone Filters available
- Compatible with many universal tips, including Optifit Tips and SafetySpace Tips
- Color coding on the pipette
- Fully autoclavable, with only 3 parts to clear
- More ergonomic design for lower required pipetting force
Spezifikation
Spezifikation
| Certifications/Compliance | ISO |
| Compatible Tips | Optifit |
| Autoclavable | Autoclavable |
| Number of Channels | 1 |
| Type | Micropipette |
| Volume (Metric) | 0.5 to 10 μL |
| Display | Clear volume |
| Filter Type | Safe-Cone |
| Finger Style | Ergonomic |
| For Use With (Application) | It is ideal for universities and colleges, or any laboratory seeking a cost-efficient liquid handling tool |
| Mehr anzeigen |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur