Learn More
Cytiva Biodyne™ Plus Membrane
Click to view available options
Dimensions (LxW):
132 x 132 mm
20 cm x 20 m
30 cm x 3 m
Unit Size:
1 roll
10 rolls
Pack of 50
Description
- Biodyne Plus membrane is recommended for detection systems that produce high background on Biodyne B membrane
Specifications
Specifications
| Packaging Type | Roll |
| Description | Non-sterile |
| Material | Positively-charged Nylon 6.6 |
| Pore Size | 0.45 μm |
| Dimensions (LxW) | 20 cm x 20 m |
| Thickness | 5.7 to 6.7 μm |