Learn More
Biodesign™ Biodesign™ Acrylic Levelling Platform
Product Code. 12866512
Product Code. 12866512
Brand: Biodesign™ G203
Only null left
Add to basket
Add to basket
Ensures a level surface when pouring and running polyacrylamide and agarose gels
10mm thick acrylic top with three individually adjustable chrome plated metal feet and a large easy to read bulls-eye level on the top.
Ensures a level surface when pouring and running polyacrylamide and agarose gels.
Specifications
Type Leveling Platform