Learn More
Corning™ BioCoat™ 96-Well, Poly-D Lysine-Treated, Flat-Bottom Microplate
Click to view available options
Color:
White
Unit Size:
Pack of 80
Description
- Superior lot-to-lot and intra-well consistency
- Minimized cross talk
- Stackable design
- Color: White
- Bottom: Clear, flat, with lid
Specifications
Specifications
| Format | Standard |
| Array | 8 x 12 |
| Packaging | 20/Pk., 80/Cs. |
| Color | White |
| Product Type | BioCoat |
| Material | Polystyrene |
| No. of Wells | 96 |
| Sterility | Sterile |
| Surface Treatment | Poly-D-Lysine |
| Well Shape | Flat Bottom (F-well) |
| Show More |