Learn More
Corning™ BioCoat™ Angiogenesis System for Endothelial Cell Invasion
GREENER_CHOICE
Product Code. 11543540
Click to view available options
Quantity:
1/Pk.
5/Pk.
Unit Size:
Each
Pack of 5
Product Code. 11543540
Brand: Corning™ 354142
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Analyze endothelial cell invasion using fluorescence detection in a simplified and reproducible manner
HTS insert plates are arrays of individual cell culture inserts connected by a rigid, robotics-friendly holder. This single-unit design makes insert plates ideal for running automated, high throughput drug transport (Caco-2 cells) cell toxicity studies or cell migration and invasion studies.
Corning BioCoat HTS Caco-2 Assay System, PET Membrane Contains specially formulated serum-free medium, culture supplements, sodium butyrate, and the Corning BioCoat Fibrillar Collagen 24-well insert system
TRUSTED_SUSTAINABILITY
Specifications
Culture Area 0.3 cm2
Material PET
Pore Size 3 μm
Type Membrane
For Use With (Application) Endothelial Cell Invasion
Quantity 5/Pk.
No. per Pack 1/Pk.
No. per Case 5/Cs.
Surface Coating Matrigel™ Matrix
No. of Wells 24