Learn More
Nitritex™ BioClean Permaflow™ Sterile Cleanroom Marker Pens
Product Code. 15298308
Click to view available options
Color:
Black
Blue
Red
Description:
Bagged
Double bagged
Double bagged, Pack of 10
Unit Size:
Pack of 10
Pack of 200
Product Code. 15298308
Brand: Nitritex™ SBPTBBK1
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Processed to ensure ISO Class 4 & EU GMP Grade A compatibility. Nitritex™ BioClean Permaflow™ Cleanroom Marker Pens are individually packaged and available with black, blue, or red ink. Double-bagged varieties are available.
  • Super permanent ink in black, blue, or red
  • Fade and water resistant
  • Quick drying
  • Non-toxic
  • Alcohol resistant labeling
  • Autoseal ink prevents drying
  • Store in a dry, cool place away from direct sunlight and fluorescent lighting
  • Manufactured in a facility holding ISO 9001:2008 certification
  • Processed in an NEBB certified ISO Class 4 cleanroom environment
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Description Bagged
Length (English) 5.5 in.
Length (Metric) 140 mm