Learn More
Ansell™ BioClean™ Notebooks Cleanroom Notebook
Product Code. 15208198
Change view
Click to view available options
Color:
Blue
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
15208198 Blue Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15208198 Supplier Ansell™ Supplier No. BNBCA5L
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Ansell™ BioClean™ Notebooks Cleanroom Notebook is the ideal choice for use in cleanrooms.
  • Polypropylene spiral and covers
  • Latex-free, low linting and heat resistant
  • Packed in an ISO Class 4 cleanroom
  • High tear and tensile strength
  • All sizes triple bagged
  • Sterilizable and autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene, Paper
Ruled Yes
Color Blue
Cover Material Low Linting Polypropylene
Certifications/Compliance Manufactured in a facility holding ISO 9001:2015 certification
Product Type Notebook
Quantity All sizes: Two books triple bagged, with each book individually double bagged.
Store in a dry, cool place (≤40°C) away from direct sunlight and fluorescent light.