Learn More
Ansell™ BioClean™ Notebooks Cleanroom Notebook
Description
- Polypropylene spiral and covers
- Latex-free, low linting and heat resistant
- Packed in an ISO Class 4 cleanroom
- High tear and tensile strength
- All sizes triple bagged
- Sterilizable and autoclavable
Specifications
Specifications
| Material | Polypropylene, Paper |
| Ruled | Yes |
| Color | Blue |
| Cover Material | Low Linting Polypropylene |
| Certifications/Compliance | Manufactured in a facility holding ISO 9001:2015 certification |
| Product Type | Notebook |
| Quantity | All sizes: Two books triple bagged, with each book individually double bagged. |
Store in a dry, cool place (≤40°C) away from direct sunlight and fluorescent light.