Learn More
Ansell BioClean Isolator Sleeve - Glove Assembly Unit C Ring and O Ring BBCO 100
Click to view available options
For Use With (Equipment):
Gloves, Sleeves
Unit Size:
Each
Description
- Enhanced convenience and reliability: Made of PVC and silicone, the BioClean™ Isolator Sleeve/Glove Assembly is specially designed to enable the easy connection of gloves to isolator sleeves
- Improved hygiene: This isolator glove assembly unit is double-bagged in EasyTear packaging, ensuring hygiene and convenience
Specifications
Specifications
| For Use With (Application) | Connecting glove to isolator sleeve |
| For Use With (Equipment) | Gloves, Sleeves |
| Product Type | O-Ring and C-Ring Set |
| Material | Silicone, PVC |
| Size | 11 |