Learn More
Ansell BioClean™ Durableu Disposable Overshoes BDBO
Description
- Resists tears, rips, and abrasions
- Low levels of particle shedding
- Resistant to a wide range of liquid chemicals
- High tensile strength
- Slip-resistant
- Non-woven spunbonded polypropylene (38 gsm)/Cast polyethylene laminate (64 gsm)
Specifications
Specifications
| Size | X-Large |
| Material | Polypropylene |
| Latex Free | No |
| Color | Blue |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Certifications/Compliance | Manufactured in a facility holding ISO 9001:2015 |
| Packaging Quantity | 50 Pieces per Sealed Inner PE Bag, 1 Inner Bag per Sealed Outer PE Bag, 8 Outer Bags per Lined Carton |
| Packaging Type | 400/Cs. |
| Product Type | Controlled Environments Shoe Covers |
Five (5) years from date of manufacture. Store in a dry, cool place (≤40°C) away from direct sunlight and fluorescent light.