missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BioBasic™ 8 PEEK Bio-Inert HPLC Columns, 5μm Particle Size

Product Code. p-4523552 Shop All Thermo Scientific Products
Click to view available options
Length (Metric):
250 mm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10620346

Brand: Thermo Scientific™ 72205252168

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Thermo Scientific™ BioBasic 8 PEEK Bio-Inert HPLC Columns, 5μm Particle Size, are optimized for the separation of a wide range of peptides.

Thermo Scientific™ BioBasic 8 PEEK Bio-Inert HPLC Columns, 5μm Particle Size, are optimized for the separation of a wide range of peptides. These high-performance columns are ideal for the analysis of proteins, peptides and other biomolecules. Optimized for the separation of medium hydrophobicity peptides, nominally less than 20,000 daltons.

  • 300Å pore size for improved biomolecule analysis
  • An excellent starting column for protein and peptide analysis
  • Outstanding reproducibility, efficiency and column lifetime
  • Excellent for LC/MS separations

Ordering Information:

Other column dimensions are available in bio-inert column hardware. Please call Customer Service for more information.

TRUSTED_SUSTAINABILITY

Specifications

Column Format PEEK Column
Diameter (Metric) 2.1 mm
For Use With LC/MS
Length (Metric) 250 mm
Particle Size 5 μm
pH 2 to 9
Pore Size 300 Å
Surface Area 100 m²/g
USP Type L7
Stationary Phase C8
Product Line BioBasic
Packing Material Silica
Carbon Load 5%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.