Learn More
Macherey-Nagel™ Blotting paper MN 218 B
Product Code. 10737811
Change view
Click to view available options
Dimensions (L x W):
210 x 90 mm
70 x 100 mm
300 x 600 mm
570 x 460 mm
580 x 600 mm
Unit Size:
Pack of 100
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) Unit Size
10737811 570 x 460 mm Pack of 100
10307981 210 x 90 mm Pack of 100
10539121 300 x 600 mm Pack of 100
10490813 580 x 600 mm Pack of 100
11922302 70 x 100 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 10737811 Supplier Macherey-Nagel™ Supplier No. 742113
In Stock
Market leading paper quality for reliable biomolecule blottings – MN 218 B
  • Speed: Slow
Spezifikation
Type Blotting Paper
Quantity 100 Pack
Thickness (Metric) 0.36 mm
Dimensions (L x W) 570 x 460 mm