Learn More
Zeiss™ Binocular Phototube
Product Code. p-8005500
Click to view available options
Type:
Binocular Phototube 20°/23 (100:0/0:100)
Binocular Phototube 30°/23 (100:0/0:100)
Binocular Phototube S 20°
Binocular Phototube, Left Port 45°/23 (50:50)
Binocular Tube 20°/23
Binocular Tube 30°/20
Unit Size:
Each
Product Code. 11845620
Brand: Zeiss™ 4255209020000
This item requires a price update prior to being ordered.
Request a Price for this item here.
Use these phototubes for a variety of applications
Binocular phototubes are available with a variety of options to suit your needs
Specifications
Type Binocular Phototube 30°/23 (100:0/0:100)
Description Reversed image with sliding prism, low-vibration prism switch; Camera port with interface 60N
Quantity 1 Ea.