missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ BIN1 (Human) Recombinant Protein
Human BIN1 full-length ORF ( AAH04101, 1 a.a. - 439 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00000274-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MAEMGSKGVTAGKIASNVQKKLTRAQEKVLQKLGKADETKDEQFEQCVQNFNKQLTEGTRLQKDLRTYLASVKAMHEASKKLNECLQEVYEPDWPGRDEANKIAENNDLLWMDYHQKLVDQALLTMDTYLGQFPDIKSRIAKRGRKLVDYDSARHHYESLQTAKKKDEAKIAKAEEELIKAQKVFEEMNVDLQEELPSLWNSRVGFYVNTFQSIAGLEENFHKEMSKLNQNLNDVLVGLEKQHGSNTFTVKAQPSDNAPAKGNKSPSPPDGSPAATPEIRVNHEPEPAGGATPGATLPKSPSQPAEASEVAGGTQPAAGAQEPGETAASEAASSSLPAVVVETFPATVNGTVEGGSGAGRLDLPPGFMFKVQAQHDYTATDTDELQLRAGDVVLVIPFQNPEEQDEGWLMGVKESDWNQHKELEKCRGVFPENFTERVP
Specifications
AAH04101 | |
bridging integrator 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BIN1 | |
GST |
274 | |
Wheat germ expression system | |
10 μg | |
AMPH2, AMPHL, DKFZp547F068, MGC10367, SH3P9 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |