Learn More
Thermo Scientific™ Bile Esculin Agar
Product Code. p-4523776
Click to view available options
Quantity:
10/Pk.
Format:
90mm Monoplate
Media Type:
Bile Esculin Agar
Manufacturing Origin:
United Kingdom
Unit Size:
Each
Product Code. 12355743
Brand: Thermo Scientific™ PO0169A
Isolate and differentiate enterococci / Group D streptococci and non-Group D streptococci using this selective microbiological medium.
Isolate and differentiate enterococci/Group D streptococci and non Group D streptococci using Thermo Scientific™ Bile Esculin Agar, a selective microbiological medium.
Enterococci / Group D streptococci hydrolyse aesculin to form aesculetin and dextrose. Aesculetin combines with ferric citrate in the medium to form a dark brown or black complex which is indicative of a positive result. Bile salts will inhibit Gram-positive bacteria other than enterococci / Group D streptococci.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need, and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications
Color Straw/ Brown, clear
Content And Storage 2°C to 10°C
Format 90mm Monoplate
Manufacturing Origin United Kingdom
Media Type Bile Esculin Agar
Product Type Agar
Fill Weight 19.5 g ± 1.0 g
Packaging Type Flow Wrapped - Single Layer
Quantity 10/Pk.