Learn More
Bilastine N-Oxide, TRC
Specifications
Specifications
| Chemical Name or Material | Bilastine N-Oxide |
| CAS | 2069238-47-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C28H37N3O4 |
| Quantity | 10 mg |
| Synonym | 1-(4-(2-Carboxypropan-2-yl)phenethyl)-4-(1-(2-ethoxyethyl)-1H-benzo[d]imidazol-2-yl)piperidine 1-Oxide |
| InChI Formula | InChI=1S/C28H37N3O4/c1-4-35-20-16-30-25-8-6-5-7-24(25)29-26(30)22-14-18-31(34,19-15-22)17-13-21-9-11-23(12-10-21)28(2,3)27(32)33/h5-12,22H,4,13-20H2,1-3H3,(H,32,33) |
| SMILES | CCOCCn1c(nc2ccccc12)C3CC[N+]([O-])(CCc4ccc(cc4)C(C)(C)C(=O)O)CC3 |
| IUPAC Name | 2-[4-[2-[4-[1-(2-ethoxyethyl)benzimidazol-2-yl]-1-oxidopiperidin-1-ium-1-yl]ethyl]phenyl]-2-methylpropanoic acid |
| Molecular Weight (g/mol) | 479.61 |
| Show More |