Description
- A mix of three different calibration beads used in the automated QC of the Bigfoot™ Spectral Cell Sorter
- Beads are retained within the Bigfoot cell sorter in a dedicated loader position
- Used during the automated QC process to verify alignment and drop delay of the system
- The onboard QC beads allow the operator to perform automated QC at any time and assist with remote support
- The Bigfoot QC bead bottle is automatically capped to minimize evaporation from the vial
- Loader location contains an integrated vortex to keep the beads suspended
- Calibration protocols are fully automated
- No pre-mixing or lot monitoring are required
- Convenient, ready to load
Specifications
Specifications
| For Use With (Equipment) | Invitrogen™ Bigfoot™ Cell Sorter |
| For Use With (Application) | Daily QC of Bigfoot Cell Sorter |
| Quantity | 3 x 5 mL |
| Product Type | QC Beads |
| Product Line | Invitrogen |