Learn More
Dostmann Electronic™ Big-digit Thermo-Hygrometer with Alarm
Product Code. 11979838
Click to view available options
Temperature Range (Metric):
-40°C to +70°C
Unit Size:
Each
Product Code. 11979838
Brand: Dostmann Electronic™ DW1DT
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Humidity Range 0 to 99%
Resolution 0.001
Temperature Range (Metric) -40°C to +70°C
Alarm Types Audible/Light
Voltage 1.5 V