missing translation for 'onlineSavingsMsg'
Learn More

Bicyclo[3.3.0]octane, Thermo Scientific™

Product Code. 17515545 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17515545 1 g 1g
17525545 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17515545 Supplier Thermo Scientific Acros Supplier No. 468520010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 694-72-4
Molecular Formula C8H14
Molecular Weight (g/mol) 110.20
MDL Number 00060882
InChI Key AEBWATHAIVJLTA-UHFFFAOYSA-N
IUPAC Name octahydropentalene
SMILES C1CC2CCCC2C1

Specifications

Alpha Vector BICYCLOOCTANE
Color Colorless
Boiling Point 137°C
Flash Point 26°C
Infrared Spectrum Conforms
Packaging Glass Bottle
Quantity 1 g
Physical Form Liquid
Chemical Name or Material Bicyclo[3.3.0]octane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.