Learn More
Tocris Bioscience™ BIBN 4096
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Potent and selective CGRP receptor antagonist (IC50 values are 0.03 and 6.4 nM for human and rat CGRP receptors respectively). Displays high affinity for human CGRP receptors (Ki = 14.4 pM); exhibits no significant affinity for 75 other receptors.
Specifications
Specifications
| CAS | 204697-65-4 |
| Quantity | 10 mg |
| Product Type | BIBN 4096 |
| Purity | >96% |
| Molecular Weight (g/mol) | 869.66 |