Learn More
Keystone Cleanroom Products BHD Blue Autoclaveable Wrapper
BIOPHARMAGREENER_CHOICE
Product Code. 17758197 Shop All Keystone Cleanroom Products Products
Click to view available options
Dimensions (L x W):
8 x 8 in.
9 x 9 in.
12 x 12 in.
15 x 15 in.
18 x 18 in.
20 x 20 in.
24 x 24 in.
30 x 30 in.
36 x 36 in.
40 x 40 in.
45 x 45 in.
45 x 48 in.
54 x 54 in.
54 x 72 in.
Unit Size:
Pack of 100
Pack of 1000
Pack of 250
Product Code. 17758197
Brand: Keystone Cleanroom Products BHDSHEET4548
When secured with autoclave tape or elasticized bands, Keystone Cleanroom Product’s Blue BHD wrappers provide an adequate microbial barrier from external sources of contamination.
  • Designed for steam sterilization
  • Treated to impart water and alcohol repellency
  • Wet formed, lightweight non-woven fabric comprised of natural wood pulp and polyester fibers bonded with a resin binder
  • Sufficient microbial barrier
  • Resists tearing and punctures
  • Does not contain any natural latex rubber
  • Processed and packaged in ISO Certified Cleanrooms
  • Certificate of Conformance included with shipment
Specifications
Autoclavable Autoclavable
Certifications/Compliance COC
Closure Type None
Color Blue
Disposable Single-use
Material Cellulose
Sterility Non-sterile
Latex Free Yes
Dimensions (L x W) 45 x 48 in.
Length (English) 48 in.
Length (Metric) 121.9 cm
Width (English) 45 in.
Width (Metric) 14.3 cm
Product Type Sterilization Wrap (Heavy-duty)