Learn More
Sartorius™ BF2 Cotton Linter Blotting Paper
Shop All Sartorius Products
Click to view available options
Length (Metric):
200 mm
460 mm
580 mm
Unit Size:
Pack of 100
Description
- Grade BF 2
- Made of high purity cotton linters for uniform buffer flow and resulting blots
- No additives to avoid any interference during the transfer
- Supplied in sheets
Specifications
Specifications
| Length (Metric) | 460 mm |
| Width (Metric) | 570 mm |
| Color | White |
| Quantity | 100/Pk. |
| Material | Cotton linters with an alpha-cellulose content |
| Thickness | 0.35 mm |