missing translation for 'onlineSavingsMsg'
Learn More

Bethanechol Chloride, TRC

Product Code. 30346684
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30346684 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30346684 Supplier Toronto Research Chemicals Supplier No. TRCB32890050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Bethanechol Chloride
CAS 590-63-6
Molecular Formula C7 H17 N2 O2 . Cl
Quantity 50 mg
Synonym Bethanechol Chloride, (2-Hydroxypropyl)trimethylammonium chloride carbamate, 1-Propanaminium, 2-[(aminocarbonyl)oxy]-N,N,N-trimethyl-, chloride, Ammonium, (2-hydroxypropyl)trimethyl-, chloride, carbamate, (±)-Bethanechol, Besacholine, Carbamylmethylcholine chloride, Mechotane, Mechothane, Mecothane, Mictone, Myocholine, Myotonachol, Myotonine chloride, NSC 30783, Trimethyl(2-carbamoyloxypropyl)ammonium chloride, Urecholine, Urecholine chloride, Uro-Carb, β-Methyl carbachol chloride, β-Methylcholine chloride urethan
InChI Formula InChI=1S/C7H16N2O2.ClH/c1-6(11-7(8)10)5-9(2,3)4;/h6H,5H2,1-4H3,(H-,8,10);1H
SMILES [Cl-].CC(C[N+](C)(C)C)OC(=O)N
IUPAC Name 2-carbamoyloxypropyl(trimethyl)azanium;chloride
Molecular Weight (g/mol) 196.68
Formula Weight 196.0979
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.