missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bethanechol Chloride, TRC
Specifications
Specifications
| Chemical Name or Material | Bethanechol Chloride |
| CAS | 590-63-6 |
| Molecular Formula | C7 H17 N2 O2 . Cl |
| Quantity | 100 mg |
| Synonym | Bethanechol Chloride, (2-Hydroxypropyl)trimethylammonium chloride carbamate, 1-Propanaminium, 2-[(aminocarbonyl)oxy]-N,N,N-trimethyl-, chloride, Ammonium, (2-hydroxypropyl)trimethyl-, chloride, carbamate, (±)-Bethanechol, Besacholine, Carbamylmethylcholine chloride, Mechotane, Mechothane, Mecothane, Mictone, Myocholine, Myotonachol, Myotonine chloride, NSC 30783, Trimethyl(2-carbamoyloxypropyl)ammonium chloride, Urecholine, Urecholine chloride, Uro-Carb, β-Methyl carbachol chloride, β-Methylcholine chloride urethan |
| InChI Formula | InChI=1S/C7H16N2O2.ClH/c1-6(11-7(8)10)5-9(2,3)4;/h6H,5H2,1-4H3,(H-,8,10);1H |
| SMILES | [Cl-].CC(C[N+](C)(C)C)OC(=O)N |
| IUPAC Name | 2-carbamoyloxypropyl(trimethyl)azanium;chloride |
| Molecular Weight (g/mol) | 196.68 |
| Formula Weight | 196.0979 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?