Learn More
Thermo Scientific™ BetaSil™ Silica Guard Cartridges
Product Code. 10187644 Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric):
1 mm I.D.
2.1 mm
3 mm
4 mm
Length (Metric):
10 mm
Particle Size:
5 μm
Unit Size:
Pack of 4
Product Code. 10187644
Brand: Thermo Scientific™ 70005012101
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Experience a powerful column for separating non-polar and moderately polar compounds based on 100Å high purity silica and designed for small molecule analysis.
  • 100Å silica
  • High surface area
TRUSTED_SUSTAINABILITY
Specifications
pH 2 to 9
Particle Size 5 μm
Length (Metric) 10 mm
Pore Size 100Å
Diameter (Metric) 2.1 mm
Diameter (Metric) Inner 2.1 mm
For Use With (Equipment) HPLC Column
Endcapped No
USP Type L3
Product Line BetaSil