Learn More
Betamethasone, TRC
Product Code. 18063320
Change view
Click to view available options
Quantity:
50 mg
100 mg
250 mg
Unit Size:
100mg
250mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18063320 50 mg 50mg
18060830 100 mg 100mg
18070740 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 18063320 Supplier Toronto Research Chemicals Supplier No. TRCB32700050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 378-44-9
Molecular Formula C22 H29 F O5
Molecular Weight (g/mol) 392.46
Synonym Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-16β-methyl- (8CI), (11β,16β)-9-Fluoro-11,17,21-trihydroxy-16-methylpregna-1,4-diene-3,20-dione, 9-Fluoro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione, 9-Fluoro-16β-methylprednisolone, 9α-Fluoro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-11β,17α,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione, 9α-Fluoro-16β-methyl-1,4-pregnadiene-11β,17α,21-triol-3,20-dione, 9α-Fluoro-16β-methylprednisolone, Becort, Bedifos, Betacorlan, Betacortil, Betacortril, Betadexamethasone, Betametasone, Betamethasone, Betamethazone, Betapredol, Betasolon, Bethamethasone, Betnelan, Betsolan, Bifas, Celestan, Celestene, Celeston, Celestone, Cidoten, Colircusi betamida, Dermabet, Desacort-Beta, Ethamethasoneb, Flosteron, Flubenisolone, NSC 39470, Rinderon, Rinderon A, Sch 4831, Visubeta, β-Methasone, β-Methasone alcohol
IUPAC Name (8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-3-one
SMILES C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C22H29FO5/c1-12-8-16-15-5-4-13-9-14(25)6-7-19(13,2)21(15,23)17(26)10-20(16,3)22(12,28)18(27)11-24/h6-7,9,12,15-17,24,26,28H,4-5,8,10-11H2,1-3H3/t12-,15-,16-,17-,19-,20-,21-,22-/m0/s1
Formula Weight 392.1999
Percent Purity >95%
Chemical Name or Material Betamethasone
Grade HPLC