Learn More
Betamethasone Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Betamethasone Acid |
| CAS | 37926-75-3 |
| Molecular Formula | C21H27FO5 |
| Quantity | 500 mg |
| Synonym | (11β,16β,17α)-9-Fluoro-11,17-dihydroxy-16-methyl-3-oxoandrosta-1,4-diene-17-carboxylic Acid, 9-Fluoro-11β,17-dihydroxy-16β-methyl-3-oxoandrosta-1,4-diene-17β-carboxylic Acid |
| InChI Formula | InChI=1S/C21H27FO5/c1-11-8-15-14-5-4-12-9-13(23)6-7-18(12,2)20(14,22)16(24)10-19(15,3)21(11,27)17(25)26/h6-7,9,11,14-16,24,27H,4-5,8,10H2,1-3H3,(H,25,26)/t11-,14-,15-,16-,18-,19-,20-,21-/m0/s1 |
| SMILES | O=C1C=C[C@@]2(C)C(CC[C@]3([H])[C@]2(F)[C@H](O)C[C@@]4(C)[C@@]3([H])C[C@H](C)[C@]4(O)C(O)=O)=C1 |
| IUPAC Name | (8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthrene-17-carboxylic acid |
| Molecular Weight (g/mol) | 378.43 |
| Formula Weight | 378.1843 |