missing translation for 'onlineSavingsMsg'
Learn More

Betamethasone 9,11-Epoxide 17-Propionate, TRC

Product Code. 30335851
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335851 5 mg 5mg
30334289 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30335851 Supplier Toronto Research Chemicals Supplier No. TRCB3270555MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Betamethasone 9,11-Epoxide 17-Propionate
CAS 79578-39-5
Molecular Formula C25 H32 O6
Quantity 5 mg
Synonym (9β,11β,16β)-9,11-Epoxy-21-hydroxy-16-methyl-17-(1-oxopropoxy)pregna-1,4-diene-3,20-dione, Beclomethasone 9,11-Epoxide 17-Propionate, Betamethasone Dipropionate EP Impurity U
InChI Formula InChI=1S/C25H32O6/c1-5-21(29)31-24(19(28)13-26)14(2)10-18-17-7-6-15-11-16(27)8-9-22(15,3)25(17)20(30-25)12-23(18,24)4/h8-9,11,14,17-18,20,26H,5-7,10,12-13H2,1-4H3/t14-,17-,18-,20-,22-,23-,24-,25+/m0/s1
SMILES CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]35O[C@H]5C[C@]12C)C(=O)CO
IUPAC Name [(1S,2S,10S,11S,13S,14R,15S,17S)-14-(2-hydroxyacetyl)-2,13,15-trimethyl-5-oxo-18-oxapentacyclo[8.8.0.01,17.02,7.011,15]octadeca-3,6-dien-14-yl] propanoate
Molecular Weight (g/mol) 428.52
Formula Weight 428.2199
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.