Learn More
Thermo Scientific™ BetaBasic™ 18 HPLC Columns, 5μm Particle Size
Available in limited dimensions. Please contact Customer Service for more details.
552.84€ - 790.18€
Specifications
Column Format | Analytical Column |
---|---|
For Use With | LC/MS, APCI |
Particle Size | 5 μm |
pH | 2 to 12 |
Pore Size | 150 Å |
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
10648425
|
Thermo Scientific™
71505-052130 |
5 μm | 2.1 mm | 50 mm |
715.32€
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||
10528985
|
Thermo Scientific™
71505-104630 |
5 μm | 4.6 mm | 100 mm |
790.18€
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||
10456684
|
Thermo Scientific™
71505-154630 |
5 μm | 4.6 mm | 150 mm |
783.05€
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||
10057454
|
Thermo Scientific™
71505-254630 |
5 μm | 4.6 mm | 250 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||
10158904
|
Thermo Scientific™
71505-032130 |
5 μm | 2.1 mm | 30 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||
10650346
|
Thermo Scientific™
71505-152130 |
5 μm | 2.1 mm | 150 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||
10418204
|
Thermo Scientific™
71505-153030 |
5 μm | 3 mm | 150 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||
10201245
|
Thermo Scientific™
71505-054630 |
5 μm | 4.6 mm | 50 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||
10169574
|
Thermo Scientific™
71505-102130 |
5 μm | 2.1 mm | 100 mm |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||
Description
Thermo ScientificTM BetaBasicTM 18 HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.
- Pore size suitable for small molecules, peptides and protein digests
- Superb pH stability
Ordering Information:
Other column dimensions are also available. Please call Customer Service for more information.
Specifications
Analytical Column | |
5 μm | |
150 Å | |
L1 | |
Silica | |
Reversed Phase | |
Reversed Phase |
LC/MS, APCI | |
2 to 12 | |
200 m²/g | |
C18 | |
13% | |
Yes |