missing translation for 'onlineSavingsMsg'
Learn More

Beta-Tocopherol (Racemic Mixture), Beta-Tocopherol (Racemic Mixture), TRC

Product Code. 30006746
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006746 5 mg 5mg
30012559 2.5 mg 2.5mg
30006842 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30006746 Supplier Toronto Research Chemicals Supplier No. TRCT5261705MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C28 H48 O2
Molecular Weight (g/mol) 416.68
Synonym 2H-1-Benzopyran-6-ol, 3,4-dihydro-2,5,8-trimethyl-2-(4,8,12-trimethyltridecyl)-, [2R*(4R*,8R*)]-, 6-Chromanol, 2,5,8-trimethyl-2-(4,8,12-trimethyltridecyl)- (8CI), rel-(2R)-3,4-Dihydro-2,5,8-trimethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-2H-1-benzopyran-6-ol, 2,5,8-Trimethyl-2-(4,8,12-trimethyltridecyl)-6-chromanol, 5,8-Dimethyltocol, Cumotocopherol, DL-β-Tocopherol, Neotocopherol, dl-β-Tocopherol, p-Xylotocopherol, β-Tocopherol
IUPAC Name (2R)-2,5,8-trimethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCc2c(C)c(O)cc(C)c2O1

Specifications

CAS 148-03-8
Quantity 5 mg
InChI Formula InChI=1S/C28H48O2/c1-20(2)11-8-12-21(3)13-9-14-22(4)15-10-17-28(7)18-16-25-24(6)26(29)19-23(5)27(25)30-28/h19-22,29H,8-18H2,1-7H3/t21-,22-,28-/m1/s1
Formula Weight 416.3654
Percent Purity >95%
Chemical Name or Material Beta Tocopherol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.