Learn More
beta-Lapachone (>90%), TRC
Specifications
Specifications
| Chemical Name or Material | Beta-Lapachone |
| CAS | 4707-32-8 |
| Molecular Formula | C15 H14 O3 |
| Quantity | 50 mg |
| Synonym | 2H-Naphtho[1,2-b]pyran-5,6-dione, 3,4-dihydro-2,2-dimethyl-, 3,4-Dihydro-2,2-dimethyl-2H-naphtho[1,2-b]pyran-5,6-dione, 2,2-Dimethyl-3,4-dihydrobenzo[h]chromene-5,6-dione, ARQ 501, MB 12066, NSC 26326, NSC 629749, SL 11001, β-Lapachone |
| InChI Formula | InChI=1S/C15H14O3/c1-15(2)8-7-11-13(17)12(16)9-5-3-4-6-10(9)14(11)18-15/h3-6H,7-8H2,1-2H3 |
| SMILES | CC1(C)CCC2=C(O1)c3ccccc3C(=O)C2=O |
| IUPAC Name | 2,2-dimethyl-3,4-dihydrobenzo[h]chromene-5,6-dione |
| Molecular Weight (g/mol) | 242.27 |
| Formula Weight | 242.0943 |