Learn More
Emma Safety Footwear Bergamo Safety Shoes
Product Code. 15998161
Change view
Click to view available options
Size:
39
40
41
42
44
45
46
47
48
Unit Size:
1 pair
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15998161 39 1 pair
15908171 40 1 pair
15918171 41 1 pair
15928171 42 1 pair
15938171 44 1 pair
15948171 45 1 pair
15958171 46 1 pair
15968171 47 1 pair
15978171 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15998161 Supplier Emma Safety Footwear Supplier No. 10709239
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Comfortable foot-hugging fit provides a naturally light and airy foot feel. Virtually penetration proof, water resistant uppers. The 100% nitrile rubber outsole repels liquids, harmful or otherwise and the ingeniously designed sole contours provide extra grip on almost all surfaces.
  • O3-Work shoe without steel toecap with sole plate
  • Full grain leather construction
  • Nitrile Rubber Heat Resistant Outsole
  • Metal-free
  • Chrome-free lining
  • ESD certified
  • Sanitized
Specifications
Certifications/Compliance EN ISO 20347:2012
Closure Type Laces
Color Black
Design Bergamo
Liner Material Hydro-Tec™ Sanitized Silver & Wellness leather
Material Leather
Size 39