Learn More
Bepotastine Isopropyl Ester, TRC
Specifications
Specifications
| Chemical Name or Material | Bepotastine Isopropyl Ester |
| CAS | 1807607-93-7 |
| Molecular Formula | C24H31ClN2O3 |
| Quantity | 25 mg |
| Synonym | 1-Methylethyl 4-[(S)-(4-chlorophenyl)-2-pyridinylmethoxy]-1-piperidinebutanoate |
| InChI Formula | InChI=1S/C24H31ClN2O3/c1-18(2)29-23(28)7-5-15-27-16-12-21(13-17-27)30-24(22-6-3-4-14-26-22)19-8-10-20(25)11-9-19/h3-4,6,8-11,14,18,21,24H,5,7,12-13,15-17H2,1-2H3/t24-/m0/s1 |
| SMILES | ClC(C=C1)=CC=C1[C@@H](C2=CC=CC=N2)OC3CCN(CCCC(OC(C)C)=O)CC3 |
| IUPAC Name | propan-2-yl 4-[4-[(S)-(4-chlorophenyl)-pyridin-2-ylmethoxy]piperidin-1-yl]butanoate |
| Molecular Weight (g/mol) | 430.97 |
| Formula Weight | 430.2023 |