Learn More
Benzyl Glycolate, TRC
Specifications
Specifications
| Chemical Name or Material | Benzyl Glycolate |
| CAS | 30379-58-9 |
| Molecular Formula | C9H10O3 |
| Quantity | 10 g |
| Synonym | Glycolic Acid Benzyl Ester (8CI), Benzyl 2-Hydroxyacetate, Benzyl glycolate, Hydroxyacetic Acid Benzyl Ester, Hydroxy Acetic Acid Phenylmethyl Ester |
| InChI Formula | InChI=1S/C9H10O3/c10-6-9(11)12-7-8-4-2-1-3-5-8/h1-5,10H,6-7H2 |
| SMILES | OCC(=O)OCc1ccccc1 |
| IUPAC Name | benzyl 2-hydroxyacetate |
| Molecular Weight (g/mol) | 166.17 |
| Formula Weight | 166.063 |