Learn More
Benzyl-d7 Alcohol, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Benzyl-d7 Alcohol |
| CAS | 71258-23-6 |
| Molecular Formula | C7 D7 H O |
| Quantity | 0.25 g |
| Synonym | Benzene-2,3,4,5,6-d5-methan-α,α-d2-ol (ACI), Benzene-d5-methan-d2-ol (9 CI), Benzyl alcohol-d7, Benzyl-d7 alcohol, Benzyl Alcohol-D7 |
| InChI Formula | InChI=1 S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8 H,6H2/i1D,2 D,3 D,4 D,5 D,6D2 |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])O |
| IUPAC Name | dideuterio-(2,3,4,5,6-pentadeuteriophenyl)methanol |
| Molecular Weight (g/mol) | 115.181 |
| Formula Weight | 115.101 g/mol |
| Show More |