missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Benzoate-d12, TRC
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
Specifications
Specifications
| Chemical Name or Material | Benzyl Benzoate-d12 |
| CAS | 352431-26-6 |
| Molecular Formula | C14 D12 O2 |
| Quantity | 2.5 mg |
| Synonym | Benzoic-d5 acid, phenyl-d5-methyl-d2 ester (9CI), Benzyl Benzoate-d12, Benzyl Benzoate-D12, Benzoic-d5 acid phenyl-d5-methyl-d2 ester, Benzoic-d5 acid benzyl-d7 ester |
| InChI Formula | InChI=1S/C14H12O2/c15-14(13-9-5-2-6-10-13)16-11-12-7-3-1-4-8-12/h1-10H,11H2/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D2 |
| SMILES | [2H]c1c([2H])c([2H])c(C(=O)OC([2H])([2H])c2c([2H])c([2H])c([2H])c([2H])c2[2H])c([2H])c1[2H] |
| IUPAC Name | [dideuterio-(2,3,4,5,6-pentadeuteriophenyl)methyl] 2,3,4,5,6-pentadeuteriobenzoate |
| Molecular Weight (g/mol) | 224.3178 |
| Formula Weight | 224.1591 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction