missing translation for 'onlineSavingsMsg'
Learn More

Benzyl Benzoate-d12, TRC

Product Code. 30335784
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335784 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30335784 Supplier Toronto Research Chemicals Supplier No. TRCB2309525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Benzyl Benzoate-d12
CAS 352431-26-6
Molecular Formula C14 D12 O2
Quantity 5 mg
Synonym Benzoic-d5 acid, phenyl-d5-methyl-d2 ester (9CI), Benzyl Benzoate-d12, Benzyl Benzoate-D12, Benzoic-d5 acid phenyl-d5-methyl-d2 ester, Benzoic-d5 acid benzyl-d7 ester
InChI Formula InChI=1S/C14H12O2/c15-14(13-9-5-2-6-10-13)16-11-12-7-3-1-4-8-12/h1-10H,11H2/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D2
SMILES [2H]c1c([2H])c([2H])c(C(=O)OC([2H])([2H])c2c([2H])c([2H])c([2H])c([2H])c2[2H])c([2H])c1[2H]
IUPAC Name [dideuterio-(2,3,4,5,6-pentadeuteriophenyl)methyl] 2,3,4,5,6-pentadeuteriobenzoate
Molecular Weight (g/mol) 224.3178
Formula Weight 224.1591
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.