missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl Benzoate-d12, CDN
Click to view available options
Quantity:
0.1 g
0.25 g
Unit Size:
0.1g
0.25g
Specifications
Specifications
| Chemical Name or Material | Benzyl Benzoate-d12 |
| CAS | 352431-26-6 |
| Molecular Formula | C14 D12 O2 |
| Quantity | 0.25 g |
| Synonym | Benzoic-d5 acid, phenyl-d5-methyl-d2 ester (9 CI), Benzyl Benzoate-d12, Benzyl Benzoate-D12, Benzoic-d5 acid phenyl-d5-methyl-d2 ester, Benzoic-d5 acid benzyl-d7 ester |
| InChI Formula | InChI=1 S/C14H12O2/c15-14(13-9-5-2-6-10-13)16-11-12-7-3-1-4-8-12/h1-10 H,11H2/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D,9 D,10 D,11D2 |
| SMILES | [2 H]c1c([2 H])c([2 H])c(C(=O)OC([2 H])([2 H])c2c([2 H])c([2 H])c([2 H])c([2 H])c2[2 H])c([2 H])c1[2 H] |
| IUPAC Name | [dideuterio-(2,3,4,5,6-pentadeuteriophenyl)methyl] 2,3,4,5,6-pentadeuteriobenzoate |
| Molecular Weight (g/mol) | 224.3178 |
| Formula Weight | 224.1591 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction