Learn More
Benzyl 2-hydroxy-2-phenylacetate, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 890-98-2 |
| Molecular Formula | C15H14O3 |
| MDL Number | MFCD00021856 |
| Quantity | 100 g |
| SMILES | O=C(OCC1=CC=CC=C1)C(O)C2=CC=CC=C2 |
| Molecular Weight (g/mol) | 242.27 |
| Percent Purity | 98% |
| Chemical Name or Material | Benzyl 2-hydroxy-2-phenylacetate |