Learn More
Benzyl 2,3-O-Isopropylidene-Alpha-D-mannofuranoside, TRC
Product Code. 30347515
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30347515 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30347515 Supplier Toronto Research Chemicals Supplier No. TRCB281200250MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Benzyl 2,3-O-Isopropylidene-a-D-mannofuranoside
CAS 20689-03-6
Molecular Formula C16 H22 O6
Quantity 250 mg
InChI Formula InChI=1S/C16H22O6/c1-16(2)21-13-12(11(18)8-17)20-15(14(13)22-16)19-9-10-6-4-3-5-7-10/h3-7,11-15,17-18H,8-9H2,1-2H3/t11-,12-,13+,14+,15+/m1/s1
SMILES CC1(C)O[C@@H]2[C@@H](OCc3ccccc3)O[C@H]([C@H](O)CO)[C@@H]2O1
IUPAC Name (1R)-1-[(3aS,4S,6R,6aS)-2,2-dimethyl-4-phenylmethoxy-3a,4,6,6a-tetrahydrofuro[3,4-d][1,3]dioxol-6-yl]ethane-1,2-diol
Molecular Weight (g/mol) 310.34
Formula Weight 310.1416